Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017762.2 | NP_060232.2 | 7 | 841 | ||
XM_005254508.2 | XP_005254565.1 | 6 | 720 | |||
XM_011521737.2 | XP_011520039.1 | 7 | 811 | |||
XM_011521738.2 | XP_011520040.1 | 7 | 811 | |||
XM_017022384.1 | XP_016877873.1 | 7 | 811 | |||
GenBank mRNA | AK000320.1 | - | 7 | 841 | ||
AK296926.1 | - | 2 | 254 | |||
AL832603.1 | - | 7 | 836 | |||
BC063296.1 | - | 6 | 749 | |||
BC094750.1 | - | 8 | 981 | |||
BC121171.1 | - | 1 | 51 | |||
BC121172.1 | - | 1 | 51 | |||
KJ534883.1 | - | 7 | 743 | |||
KJ535030.1 | - | 7 | 749 | |||
KJ535071.1 | - | 6 | 658 | |||
KJ535075.1 | - | 7 | 742 | |||
KJ535083.1 | - | 6 | 618 |