Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017762.2 | NP_060232.2 | 10, 11 | 1159 | ||
XM_005254508.2 | XP_005254565.1 | 9, 10 | 1038 | |||
XM_011521737.2 | XP_011520039.1 | 10, 11 | 1129 | |||
XM_011521738.2 | XP_011520040.1 | 10, 11 | 1129 | |||
XM_017022384.1 | XP_016877873.1 | 10, 11 | 1129 | |||
GenBank mRNA | AK000320.1 | - | 10, 11 | 1159 | ||
AK296926.1 | - | 5, 6 | 572 | |||
AL832603.1 | - | 10, 11 | 1154 | |||
BC063296.1 | - | 9, 10 | 1067 | |||
BC094750.1 | - | 11, 12 | 1299 | |||
BC121171.1 | - | 4, 5 | 369 | |||
BC121172.1 | - | 4, 5 | 369 | |||
KJ534883.1 | - | 10, 11 | 1061 | |||
KJ535030.1 | - | 10, 11 | 1067 | |||
KJ535071.1 | - | 9, 10 | 976 | |||
KJ535075.1 | - | 10, 11 | 1060 | |||
KJ535083.1 | - | 9, 10 | 936 |