Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271420.1 | NP_001258349.1 | 10 | 1829 | ||
NM_017775.3 | NP_060245.3 | 10 | 1692 | |||
GenBank mRNA | AK000350.1 | - | 8 | 908 | ||
AK025958.1 | - | 4 | 578 | |||
AK055780.1 | - | 10 | 1385 | |||
AK056878.1 | - | 10 | 1604 | |||
AK094819.1 | - | 3 | 1886 | |||
AK297783.1 | - | 10 | 1248 | |||
AL832412.1 | - | 2 | 1617 | |||
BC011698.2 | - | 10 | 1231 | |||
BC073796.1 | - | 10 | 1235 | |||
BC105128.1 | - | 10 | 1692 | 8144131 | ||
BC112107.1 | - | 10 | 1692 | 8327575 |