Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317784.1 | NP_001304713.1 | 3 | 558 | ||
NM_017988.5 | NP_060458.3 | 3 | 710 | |||
XM_017019618.1 | XP_016875107.1 | 3 | 558 | |||
XM_017019619.1 | XP_016875108.1 | 3 | 710 | |||
XM_017019620.1 | XP_016875109.1 | 3 | 561 | |||
GenBank mRNA | AK000936.1 | - | 2 | 316 | ||
AK125091.1 | - | 3 | 695 | |||
AK292980.1 | - | 3 | 484 | |||
BC063798.1 | - | 3 | 323 | 6473586 | ||
DA234206.1 | - | 3 | 558 |