Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317784.1 | NP_001304713.1 | 18 | 2855 | ||
NM_017988.5 | NP_060458.3 | 18 | 3007 | |||
XM_017019618.1 | XP_016875107.1 | 19 | 2867 | |||
XM_017019619.1 | XP_016875108.1 | 19 | 3019 | |||
XM_017019620.1 | XP_016875109.1 | 20 | 3006 | |||
GenBank mRNA | AB037781.1 | - | 16 | 2173 | ||
AK001597.1 | - | 8 | 1167 | |||
AK024274.1 | - | 14 | 2016 | |||
AK026160.1 | - | 1 | 272 | |||
AK027551.1 | - | 14 | 2073 | |||
AK292980.1 | - | 18 | 2781 | |||
BC012387.2 | - | 9 | 1300 | |||
BC063798.1 | - | 18 | 2620 | 6473586 |