Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144856.1 | NP_001138328.1 | 5 | 757 | ||
NM_001144857.1 | NP_001138329.1 | 4 | 925 | |||
NM_018173.3 | NP_060643.2 | 5 | 675 | |||
XM_005253704.4 | XP_005253761.1 | 5 | 772 | |||
XM_006718985.3 | XP_006719048.1 | 5 | 772 | |||
XM_011520967.2 | XP_011519269.1 | 5 | 647 | |||
XM_017019555.1 | XP_016875044.1 | 5 | 772 | |||
GenBank mRNA | AK001527.1 | - | 5 | 676 | ||
AK023741.1 | - | 5 | 757 | |||
AK095373.1 | - | 4 | 925 | |||
BC104751.1 | - | 5 | 611 | 40035012 | ||
BC105961.1 | - | 5 | 611 | 40035014 | ||
BC109095.2 | - | 5 | 611 | 40034695 |