Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320613.1 | NP_001307542.1 | 2 | 1056 | ||
NM_004036.4 | NP_004027.2 | 2 | 1056 | |||
XM_006711925.1 | XP_006711988.1 | 2 | 772 | |||
XM_011532489.1 | XP_011530791.1 | 2 | 772 | |||
XM_011532490.1 | XP_011530792.1 | 2 | 772 | |||
XM_011532491.1 | XP_011530793.1 | 2 | 772 | |||
XM_011532492.2 | XP_011530794.1 | 2 | 772 | |||
XM_011532493.1 | XP_011530795.1 | 2 | 772 | |||
XM_011532494.1 | XP_011530796.1 | 2 | 772 | |||
XM_017003186.1 | XP_016858675.1 | 2 | 772 | |||
XM_017003187.1 | XP_016858676.1 | 2 | 772 | |||
XM_017003188.1 | XP_016858677.1 | 2 | 772 | |||
XM_017003189.1 | XP_016858678.1 | 2 | 772 | |||
XM_017003190.1 | XP_016858679.1 | 2 | 772 | |||
GenBank mRNA | AF033861.1 | - | 1 | 671 | ||
AK027857.1 | - | 1 | 567 | |||
AK122926.1 | - | 2 | 654 | |||
BC126235.1 | - | 1 | 582 | 8991951 | ||
BC144119.1 | - | 1 | 582 | 9052639 |