Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020198.2 | NP_064583.2 | 3 | 698 | ||
XM_005257527.2 | XP_005257584.1 | 3 | 566 | |||
GenBank mRNA | AF113221.1 | - | 1 | 44 | ||
AF226054.1 | - | 3 | 546 | |||
AK027786.1 | - | 3 | 540 | |||
AK027844.1 | - | 3 | 527 | |||
AK075387.1 | - | 4 | 627 | |||
AK225813.1 | - | 3 | 568 | |||
AK314328.1 | - | 3 | 553 | |||
BC008905.2 | - | 3 | 520 | 3688068 |