Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144955.1 | NP_001138427.1 | 3 | 525 | ||
NM_020234.5 | NP_064619.2 | 4 | 653 | |||
XM_011521815.1 | XP_011520117.1 | 3 | 574 | |||
XM_017022419.1 | XP_016877908.1 | 3 | 572 | |||
XM_017022420.1 | XP_016877909.1 | 4 | 1434 | |||
XM_017022421.1 | XP_016877910.1 | 4 | 665 | |||
XM_017022422.1 | XP_016877911.1 | 3 | 1306 | |||
XM_017022423.1 | XP_016877912.1 | 3 | 574 | |||
XM_017022424.1 | XP_016877913.1 | 4 | 645 | |||
XM_017022425.1 | XP_016877914.1 | 4 | 645 | |||
XM_017022426.1 | XP_016877915.1 | 3 | 1306 | |||
XM_017022427.1 | XP_016877916.1 | 4 | 590 | |||
XM_017022428.1 | XP_016877917.1 | 4 | 595 | |||
XM_017022429.1 | XP_016877918.1 | 4 | 1541 | |||
XM_017022430.1 | XP_016877919.1 | 4 | 744 | |||
XM_017022431.1 | XP_016877920.1 | 3 | 186 | |||
GenBank mRNA | AF168717.1 | - | 3 | 502 | ||
AK290184.1 | - | 3 | 494 | |||
BC032535.2 | - | 4 | 593 | 5527713 | ||
BC093073.1 | - | 4 | 583 | 30326731 |