Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144955.1 | NP_001138427.1 | 4 | 587 | ||
NM_020234.5 | NP_064619.2 | 5 | 715 | |||
XM_011521815.1 | XP_011520117.1 | 4 | 636 | |||
XM_017022419.1 | XP_016877908.1 | 4 | 634 | |||
XM_017022420.1 | XP_016877909.1 | 5 | 1496 | |||
XM_017022421.1 | XP_016877910.1 | 5 | 727 | |||
XM_017022422.1 | XP_016877911.1 | 4 | 1368 | |||
XM_017022423.1 | XP_016877912.1 | 4 | 636 | |||
XM_017022424.1 | XP_016877913.1 | 5 | 707 | |||
XM_017022425.1 | XP_016877914.1 | 5 | 707 | |||
XM_017022426.1 | XP_016877915.1 | 4 | 1368 | |||
XM_017022427.1 | XP_016877916.1 | 5 | 652 | |||
XM_017022428.1 | XP_016877917.1 | 5 | 657 | |||
XM_017022429.1 | XP_016877918.1 | 5 | 1603 | |||
XM_017022430.1 | XP_016877919.1 | 5 | 806 | |||
XM_017022431.1 | XP_016877920.1 | 4 | 248 | |||
GenBank mRNA | AF168717.1 | - | 4 | 565 | ||
AK290184.1 | - | 4 | 556 | |||
BC032535.2 | - | 5 | 655 | 5527713 | ||
BC093073.1 | - | 5 | 645 | 30326731 |
Gene Symbol: NMNAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320510.1 | NP_001307439.1 | 4 | 648 | ||
XM_017006328.1 | XP_016861817.1 | 5 | 665 | |||
XM_017006329.1 | XP_016861818.1 | 5 | 662 | |||
XM_017006330.1 | XP_016861819.1 | 4 | 625 | |||
XM_017006331.1 | XP_016861820.1 | 5 | 738 | |||
XM_017006332.1 | XP_016861821.1 | 5 | 4679 | |||
XM_017006333.1 | XP_016861822.1 | 3 | 528 | |||
XM_017006334.1 | XP_016861823.1 | 3 | 525 | |||
XM_017006339.1 | XP_016861828.1 | 4 | 563 | |||
XM_017006340.1 | XP_016861829.1 | 3 | 532 |