Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300743.1 | NP_001287672.1 | 4 | 627 | ||
NM_020314.5 | NP_064710.4 | 4 | 627 | |||
XM_005255435.3 | XP_005255492.1 | 4 | 579 | |||
XM_006721065.3 | XP_006721128.1 | 4 | 579 | |||
XM_011545896.2 | XP_011544198.1 | 3 | 257 | |||
GenBank mRNA | AK095078.1 | - | 4 | 324 | ||
AK290286.1 | - | 4 | 324 | |||
AK294885.1 | - | 4 | 627 | |||
AL832159.1 | - | 4 | 323 | |||
AL833428.1 | - | 4 | 316 | |||
BC014900.2 | - | 3 | 295 | 3909068 | ||
BC050464.1 | - | 4 | 367 | 6061139 | ||
BC058845.1 | - | 4 | 326 | 6452778 | ||
BX537867.1 | - | 4 | 339 |