Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LYRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128301.2 | NP_001121773.1 | 5 | 1427 | ||
NM_001128302.2 | NP_001121774.1 | 4 | 1195 | |||
NM_001302835.1 | NP_001289764.1 | 6 | 1371 | |||
NM_001302836.1 | NP_001289765.1 | 6 | 1523 | |||
NM_020424.4 | NP_065157.1 | 5 | 1346 | |||
NR_126516.1 | 4 | 1268 | ||||
NR_126520.1 | 3 | 1050 | ||||
NR_126521.1 | 3 | 1036 | ||||
NR_126527.1 | 3 | 864 | ||||
NR_126528.1 | 4 | 1282 | ||||
XM_005255444.1 | XP_005255501.1 | 5 | 1248 | |||
XM_011545900.2 | XP_011544202.1 | 5 | 1010 | |||
XM_011545901.2 | XP_011544203.1 | 5 | 1076 | |||
XM_011545902.1 | XP_011544204.1 | 6 | 1347 | |||
XM_011545908.2 | XP_011544210.1 | 5 | 1078 | |||
XM_017023475.1 | XP_016878964.1 | 6 | 1763 | |||
XM_017023476.1 | XP_016878965.1 | 4 | 1661 | |||
XM_017023477.1 | XP_016878966.1 | 4 | 1598 | |||
XM_017023478.1 | XP_016878967.1 | 5 | 1068 | |||
XM_017023479.1 | XP_016878968.1 | 5 | 1505 | |||
XM_017023480.1 | XP_016878969.1 | 7 | 1591 | |||
XM_017023481.1 | XP_016878970.1 | 5 | 1220 | |||
XM_017023482.1 | XP_016878971.1 | 4 | 1274 | |||
GenBank mRNA | AK308690.1 | - | 3 | 1116 | ||
AL359616.1 | - | 1 | 448 | |||
BC017039.1 | - | 6 | 1372 | 3849248 |