Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301036.1 | NP_001287965.1 | 4 | 2129 | ||
NM_020698.3 | NP_065749.2 | 4 | 2110 | |||
GenBank mRNA | AB032971.1 | - | 4 | 2039 | ||
AB073655.1 | - | 1 | 605 | |||
BC040535.1 | - | 4 | 2096 | 5264859 |