Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020774.3 | NP_065825.1 | 17 | 2707 | ||
XM_011526098.1 | XP_011524400.1 | 9 | 1708 | |||
XM_017025873.1 | XP_016881362.1 | 14 | 2399 | |||
GenBank mRNA | AB037744.1 | - | 5 | 623 | ||
AK075157.1 | - | 5 | 574 | |||
AK091266.1 | - | 16 | 2137 | |||
AL713705.1 | - | 2 | 136 | |||
AY147849.1 | - | 17 | 2725 | |||
AY149908.1 | - | 17 | 2452 | |||
BC022403.1 | - | 7 | 828 | |||
BC110581.1 | - | 17 | 2452 | 40008394 | ||
BC110582.1 | - | 17 | 2452 | 40008395 | ||
CR749635.1 | - | 14 | 1872 |