Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004064.4 | NP_004055.1 | 1 | 1026 | ||
GenBank mRNA | AB451291.1 | - | 1 | 456 | ||
AB451423.1 | - | 1 | 456 | |||
AF247551.1 | - | 1 | 456 | |||
AK298335.1 | - | 2 | 545 | |||
AK312461.1 | - | 1 | 931 | |||
AY004255.1 | - | 1 | 608 | |||
BC001971.1 | - | 1 | 833 | 3458141 | ||
BT019553.1 | - | 1 | 456 | |||
BT019554.1 | - | 1 | 456 | |||
CR457399.1 | - | 1 | 456 | |||
U10906.1 | - | 1 | 456 |