Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 12 | 1748 | ||
NM_001258373.1 | NP_001245302.1 | 13 | 1851 | |||
NM_022827.3 | NP_073738.2 | 13 | 1796 | |||
GenBank mRNA | AK025622.1 | - | 8 | 1037 | ||
AK092731.1 | - | 13 | 1781 | |||
AK125807.1 | - | 14 | 2618 | |||
AK291805.1 | - | 13 | 1739 | |||
BC017468.1 | - | 2 | 240 | |||
BC025255.1 | - | 13 | 1684 | 5019010 | ||
BC065526.1 | - | 13 | 1738 | 6149633 | ||
BC111029.1 | - | 12 | 1653 | 5245538 | ||
CR936769.1 | - | 11 | 1986 | |||
GQ472220.1 | - | 12 | 1653 |