Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282683.1 | NP_001269612.1 | 10 | 3341 | ||
NM_024735.4 | NP_079011.3 | 9 | 3593 | |||
GenBank mRNA | AF318348.1 | - | 8 | 3791 | ||
AF428140.1 | - | 9 | 3557 | |||
AL117444.1 | - | 7 | 3101 | |||
BC012748.1 | - | 9 | 3567 | 3958783 | ||
BI914636.1 | - | 1 | 30 |