Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025082.3 | NP_079358.3 | 14 | 1939 | ||
XM_006721277.2 | XP_006721340.1 | 12 | 1794 | |||
XM_011523344.1 | XP_011521646.1 | 13 | 1916 | |||
XM_011523345.1 | XP_011521647.1 | 5 | 917 | |||
XM_017023712.1 | XP_016879201.1 | 12 | 1793 | |||
XM_017023713.1 | XP_016879202.1 | 12 | 2221 | |||
XM_017023714.1 | XP_016879203.1 | 12 | 2326 | |||
XM_017023715.1 | XP_016879204.1 | 9 | 1666 | |||
XM_017023716.1 | XP_016879205.1 | 12 | 2587 | |||
XM_017023717.1 | XP_016879206.1 | 5 | 1029 | |||
GenBank mRNA | AK023173.1 | - | 10 | 1729 | ||
AK056097.1 | - | 14 | 1939 | |||
AK225399.1 | - | 11 | 1428 | |||
AK291527.1 | - | 11 | 1459 | |||
AK300887.1 | - | 9 | 1605 | |||
AK301621.1 | - | 11 | 1342 | |||
BC007642.1 | - | 11 | 1382 | |||
BC007864.2 | - | 2 | 188 | |||
BC015202.2 | - | 11 | 1390 | 3878214 | ||
BX648617.1 | - | 7 | 1369 |