Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142448.1 | NP_001135920.1 | 13 | 1938 | ||
NM_001142449.1 | NP_001135921.1 | 10 | 1953 | |||
NM_001142450.1 | NP_001135922.1 | 13 | 2088 | |||
NM_001142451.1 | NP_001135923.1 | 11 | 2019 | |||
NM_032038.2 | NP_114427.1 | 12 | 2175 | |||
XM_006721096.3 | XP_006721159.1 | 12 | 1789 | |||
XM_017023758.1 | XP_016879247.1 | 13 | 1957 | |||
XM_017023759.1 | XP_016879248.1 | 13 | 1941 | |||
GenBank mRNA | AF212371.1 | - | 12 | 2122 | ||
AF370423.1 | - | 13 | 2069 | |||
AL390215.1 | - | 10 | 1915 | |||
BC006156.1 | - | 12 | 1786 | |||
BC008325.1 | - | 12 | 2000 | 3501826 | ||
BC038961.1 | - | 13 | 1952 | 6053095 | ||
BC047741.1 | - | 11 | 1993 | 5757339 | ||
BC065235.1 | - | 13 | 1920 | 6161778 |