Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032206.4 | NP_115582.4 | 5 | 1246 | ||
XM_005256193.2 | XP_005256250.1 | 6 | 1307 | |||
XM_005256194.3 | XP_005256251.1 | 6 | 1518 | |||
XM_005256195.2 | XP_005256252.1 | 6 | 1307 | |||
XM_005256196.2 | XP_005256253.1 | 6 | 1307 | |||
XM_005256197.2 | XP_005256254.1 | 6 | 1307 | |||
XM_005256199.3 | XP_005256256.1 | 6 | 1307 | |||
XM_005256201.3 | XP_005256258.1 | 6 | 1307 | |||
XM_006721297.2 | XP_006721360.1 | 6 | 1295 | |||
XM_006721298.2 | XP_006721361.1 | 6 | 1307 | |||
XM_006721300.2 | XP_006721363.1 | 6 | 1307 | |||
XM_011523373.2 | XP_011521675.1 | 6 | 1454 | |||
XM_011523375.2 | XP_011521677.1 | 6 | 1307 | |||
XM_017023766.1 | XP_016879255.1 | 6 | 1307 | |||
XM_017023767.1 | XP_016879256.1 | 6 | 1295 | |||
XM_017023768.1 | XP_016879257.1 | 6 | 1307 | |||
XM_017023769.1 | XP_016879258.1 | 6 | 1295 | |||
XM_017023770.1 | XP_016879259.1 | 6 | 1307 | |||
XM_017023771.1 | XP_016879260.1 | 6 | 1307 | |||
GenBank mRNA | AF389420.1 | - | 4 | 1119 | ||
AK025362.1 | - | 1 | 378 | |||
AK074182.1 | - | 5 | 1259 | |||
AK090439.1 | - | 1 | 301 | |||
AK097030.1 | - | 6 | 1296 | |||
EF452236.1 | - | 4 | 1119 |