Search Thermo Fisher Scientific
Gene Symbol: TMEM117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286211.1 | NP_001273140.1 | 7 | 2433 | ||
NM_001286212.1 | NP_001273141.1 | 7 | 2463 | |||
NM_001286213.1 | NP_001273142.1 | 7 | 2430 | |||
NM_032256.2 | NP_115632.1 | 8 | 2563 | |||
XM_011538831.2 | XP_011537133.1 | 8 | 2572 | |||
XM_011538832.2 | XP_011537134.1 | 8 | 2247 | |||
XM_011538833.2 | XP_011537135.1 | 7 | 2064 | |||
XM_017020022.1 | XP_016875511.1 | 8 | 2154 | |||
XM_017020023.1 | XP_016875512.1 | 9 | 2395 | |||
GenBank mRNA | AK057287.1 | - | 8 | 2282 | ||
AK097511.1 | - | 3 | 1507 | |||
AL136855.1 | - | 8 | 2265 | |||
BC030284.1 | - | 7 | 2171 | 5240522 | ||
BC060798.1 | - | 8 | 2326 | 30348706 |