Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143852.1 | NP_001137324.1 | 13 | 2680 | ||
NM_032300.4 | NP_115676.1 | 13 | 2741 | |||
XM_011538836.2 | XP_011537138.1 | 13 | 3016 | |||
XM_011538837.2 | XP_011537139.1 | 13 | 2589 | |||
XM_017020025.1 | XP_016875514.1 | 13 | 2925 | |||
GenBank mRNA | AK000079.1 | - | 1 | 126 | ||
AY007230.1 | - | 13 | 2742 | |||
BX648596.1 | - | 2 | 2404 |