Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004139.4 | NP_004130.2 | 3 | 465 | ||
GenBank mRNA | AF105067.1 | - | 3 | 376 | ||
AK313625.1 | - | 3 | 395 | |||
AY344471.1 | - | 3 | 382 | |||
BC022256.1 | - | 3 | 394 | 4767724 | ||
M35533.1 | - | 3 | 359 |