Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184906.1 | NP_001171835.1 | 8 | 800 | ||
NM_032875.2 | NP_116264.2 | 9 | 896 | |||
XM_005257746.3 | XP_005257803.2 | 9 | 1087 | |||
XM_005257747.3 | XP_005257804.2 | 8 | 868 | |||
GenBank mRNA | AK123048.1 | - | 9 | 897 | ||
AK291844.1 | - | 9 | 907 | |||
AK312612.1 | - | 9 | 877 | |||
BC007557.1 | - | 9 | 863 | 2987858 | ||
DQ223959.1 | - | 8 | 767 |