Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040153.3 | NP_001035243.2 | 6 | 1232 | ||
NM_001242868.1 | NP_001229797.1 | 6 | 1741 | |||
NM_001242869.1 | NP_001229798.1 | 5 | 1004 | |||
NM_001242870.1 | NP_001229799.1 | 4 | 854 | |||
NM_001242871.1 | NP_001229800.1 | 5 | 1280 | |||
NM_144595.3 | NP_653196.1 | 5 | 1196 | |||
XM_011534929.2 | XP_011533231.1 | 7 | 1621 | |||
XM_011534930.1 | XP_011533232.1 | 6 | 1134 | |||
GenBank mRNA | AK054608.1 | - | 5 | 976 | ||
AK094337.1 | - | 1 | 777 | |||
AK289600.1 | - | 5 | 831 | |||
AK289714.1 | - | 5 | 986 | |||
AK294201.1 | - | 6 | 1232 | |||
AK295423.1 | - | 5 | 1056 | |||
AL834203.1 | - | 4 | 718 | |||
AL834332.1 | - | 5 | 989 | |||
BC045177.1 | - | 5 | 1005 | |||
BC111497.1 | - | 5 | 987 | 6148614 |