Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166695.2 | NP_001160167.1 | 2 | 598 | ||
NM_001166696.2 | NP_001160168.1 | 2 | 598 | |||
NM_001289939.1 | NP_001276868.1 | 2 | 598 | |||
NM_001289940.1 | NP_001276869.1 | 2 | 598 | |||
NM_004172.4 | NP_004163.3 | 2 | 598 | |||
XM_005248342.2 | XP_005248399.1 | 3 | 451 | |||
XM_011514084.2 | XP_011512386.1 | 2 | 347 | |||
GenBank mRNA | AK057823.1 | - | 2 | 347 | ||
AK293886.1 | - | 2 | 342 | |||
AK295754.1 | - | 2 | 336 | |||
AK312304.1 | - | 2 | 366 | |||
AY954110.1 | - | 1 | 122 | |||
BC037310.2 | - | 2 | 366 | 5264000 | ||
BC054475.1 | - | 2 | 346 | |||
D26443.1 | - | 2 | 300 | |||
L19158.1 | - | 2 | 261 | |||
U03504.1 | - | 1 | 152 |