Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320321.1 | NP_001307250.1 | 8 | 2000 | ||
NM_152588.2 | NP_689801.1 | 9 | 2571 | |||
XM_017018884.1 | XP_016874373.1 | 10 | 2918 | |||
GenBank mRNA | AK074634.1 | - | 7 | 865 | ||
AK299771.1 | - | 8 | 1674 | |||
AK315285.1 | - | 9 | 2215 | |||
AL834509.1 | - | 9 | 2571 | |||
AX795392.1 | - | 9 | 2846 | |||
AX795394.1 | - | 10 | 3923 | |||
BC093852.1 | - | 9 | 2228 | 7939697 | ||
BC093854.1 | - | 9 | 2228 | 7939699 | ||
BC143378.1 | - | 8 | 1657 |