Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKS1B SIRNA Assays ›
Gene Symbol:
ANKS1B
Gene Name:
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases:
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location:
Chr.12: 98729904 - 99984727 on Build GRCh38
UniGene ID:
Hs.506458
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141560
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKS1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204065.1
NP_001190994.1
2
309
NM_001204067.1
NP_001190996.1
5
522
NM_001204068.1
NP_001190997.1
4
805
NM_001204069.1
NP_001190998.1
4
805
NM_001204070.1
NP_001190999.1
4
805
NM_001204079.1
NP_001191008.1
2
309
NM_001204080.1
NP_001191009.1
2
309
NM_001204081.1
NP_001191010.1
2
309
NM_020140.3
NP_064525.1
4
805
NM_152788.4
NP_690001.3
18
3237
NM_181670.3
NP_858056.2
4
805
XM_005269028.4
XP_005269085.1
18
2848
XM_005269029.4
XP_005269086.1
18
2808
XM_005269032.3
XP_005269089.1
2
173
XM_006719504.3
XP_006719567.1
18
3310
XM_006719505.3
XP_006719568.1
18
3310
XM_006719506.3
XP_006719569.1
18
3310
XM_006719507.3
XP_006719570.1
18
3310
XM_006719508.3
XP_006719571.1
18
3310
XM_006719509.3
XP_006719572.1
18
3310
XM_006719510.3
XP_006719573.1
18
3310
XM_006719512.3
XP_006719575.1
18
3310
XM_006719513.3
XP_006719576.1
18
3310
XM_006719514.3
XP_006719577.1
18
3298
XM_011538571.2
XP_011536873.1
18
3310
XM_017019651.1
XP_016875140.1
18
3310
XM_017019652.1
XP_016875141.1
18
3298
XM_017019653.1
XP_016875142.1
18
3310
XM_017019654.1
XP_016875143.1
18
3298
XM_017019655.1
XP_016875144.1
18
3310
XM_017019656.1
XP_016875145.1
18
3310
XM_017019657.1
XP_016875146.1
18
3310
XM_017019658.1
XP_016875147.1
18
3310
XM_017019659.1
XP_016875148.1
4
801
XM_017019660.1
XP_016875149.1
4
801
XM_017019661.1
XP_016875150.1
5
515
XM_017019662.1
XP_016875151.1
5
645
XM_017019663.1
XP_016875152.1
4
801
XM_017019664.1
XP_016875153.1
5
457
XM_017019665.1
XP_016875154.1
4
801
GenBank mRNA
AF145204.1
-
12
1941
AF164792.1
-
4
738
AK289768.1
-
4
770
AK294191.1
-
5
522
AK294994.1
-
2
181
AK295988.1
-
2
282
AK315954.1
-
2
164
AM980945.1
-
2
165
AY281131.1
-
18
2823
AY281132.1
-
4
498
AY283057.2
-
11
1854
AY356353.1
-
2
138
AY620824.1
-
9
1548
AY753193.1
-
4
498
BC026313.1
-
4
700
BC142669.1
-
4
752
BC150204.1
-
4
752
BP193713.1
-
2
281
DB475766.1
-
2
310
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of synaptic plasticity by receptor localization to synapse
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0