Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPECC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033553.2 | NP_001028725.1 | 4 | 717 | ||
NM_001033554.2 | NP_001028726.1 | 2 | 626 | |||
NM_001033555.2 | NP_001028727.1 | 2 | 626 | |||
NM_001243438.1 | NP_001230367.1 | 2 | 626 | |||
NM_001243439.1 | NP_001230368.1 | 4 | 776 | |||
NM_152904.4 | NP_690868.3 | 4 | 717 | |||
XM_011524075.1 | XP_011522377.1 | 3 | 669 | |||
XM_017025317.1 | XP_016880806.1 | 3 | 669 | |||
XM_017025318.1 | XP_016880807.1 | 3 | 669 | |||
XM_017025319.1 | XP_016880808.1 | 4 | 811 | |||
XM_017025320.1 | XP_016880809.1 | 4 | 811 | |||
XM_017025321.1 | XP_016880810.1 | 2 | 626 | |||
XM_017025322.1 | XP_016880811.1 | 3 | 669 | |||
XM_017025323.1 | XP_016880812.1 | 3 | 669 | |||
XM_017025324.1 | XP_016880813.1 | 3 | 669 | |||
XM_017025325.1 | XP_016880814.1 | 4 | 811 | |||
GenBank mRNA | AB041533.1 | - | 4 | 717 | ||
AK295093.1 | - | 4 | 749 | |||
AK295811.1 | - | 1 | 253 | |||
AK304186.1 | - | 2 | 548 | |||
AK314341.1 | - | 4 | 792 | |||
AY816326.1 | - | 2 | 531 | |||
AY816327.1 | - | 2 | 531 | |||
AY816328.1 | - | 4 | 717 | |||
AY816329.1 | - | 4 | 717 | |||
BC021123.1 | - | 2 | 524 | 3614358 | ||
BC050058.1 | - | 2 | 461 | 5277450 | ||
BP287096.1 | - | 1 | 89 | |||
BP309862.1 | - | 1 | 279 |