Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153228.2 | NP_694960.2 | 5 | 451 | ||
XM_006721728.3 | XP_006721791.1 | 4 | 674 | |||
XM_011524428.2 | XP_011522730.1 | 5 | 451 | |||
XM_011524429.2 | XP_011522731.1 | 5 | 608 | |||
XM_011524430.2 | XP_011522732.1 | 6 | 574 | |||
XM_011524431.2 | XP_011522733.1 | 5 | 506 | |||
XM_017024263.1 | XP_016879752.1 | 7 | 668 | |||
XM_017024264.1 | XP_016879753.1 | 7 | 1653 | |||
XM_017024265.1 | XP_016879754.1 | 7 | 862 | |||
XM_017024266.1 | XP_016879755.1 | 5 | 623 | |||
XM_017024267.1 | XP_016879756.1 | 4 | 475 | |||
XM_017024268.1 | XP_016879757.1 | 6 | 1559 | |||
XM_017024269.1 | XP_016879758.1 | 4 | 748 | |||
XM_017024270.1 | XP_016879759.1 | 7 | 666 | |||
GenBank mRNA | AK095654.1 | - | 5 | 451 |