Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEMA6D SIRNA Assays ›
Gene Symbol:
SEMA6D
Gene Name:
semaphorin 6D
Gene Aliases:
-
Chromosome Location:
Chr.15: 47184101 - 47774223 on Build GRCh38
UniGene ID:
Hs.511265
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141631
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEMA6D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198999.1
NP_001185928.1
11
1040
NM_020858.1
NP_065909.1
8
1028
NM_024966.2
NP_079242.2
8
1028
NM_153616.1
NP_705869.1
8
1028
NM_153617.1
NP_705870.1
8
1028
NM_153618.1
NP_705871.1
8
1028
NM_153619.1
NP_705872.1
8
1028
XM_005254685.3
XP_005254742.1
8
990
XM_005254686.2
XP_005254743.1
8
934
XM_005254687.2
XP_005254744.1
8
934
XM_005254689.3
XP_005254746.1
8
990
XM_011522075.2
XP_011520377.1
11
1145
XM_011522076.2
XP_011520378.1
8
934
XM_011522077.2
XP_011520379.1
8
934
XM_011522078.2
XP_011520380.1
8
934
XM_011522079.2
XP_011520381.1
8
934
XM_011522080.2
XP_011520382.1
8
934
XM_011522081.2
XP_011520383.1
8
934
XM_017022617.1
XP_016878106.1
11
958
XM_017022618.1
XP_016878107.1
8
990
XM_017022619.1
XP_016878108.1
8
990
XM_017022620.1
XP_016878109.1
11
1145
XM_017022621.1
XP_016878110.1
11
1145
GenBank mRNA
AB040912.2
-
11
1040
AF389426.1
-
8
1028
AF389427.1
-
8
1028
AF389428.1
-
8
1028
AF389429.1
-
8
1028
AF389430.1
-
8
1028
AK021660.1
-
5
326
BC150253.1
-
11
1040
9007179
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane-bound signaling molecule
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
neural crest cell migration
positive regulation of smooth muscle cell migration
negative regulation of smooth muscle cell migration
ventricular system development
positive regulation of cell migration
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
semaphorin receptor binding
chemorepellent activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0