Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135919.1 | NP_001129391.1 | 3 | 1129 | ||
NM_181785.3 | NP_861450.1 | 3 | 1129 | |||
XM_005266361.2 | XP_005266418.1 | 3 | 807 | |||
GenBank mRNA | AF315594.1 | - | 3 | 713 | ||
AK093932.1 | - | 2 | 1557 | |||
AK098426.1 | - | 1 | 159 | |||
AK124604.1 | - | 1 | 1821 | |||
BC036662.1 | - | 3 | 1091 | 4827143 | ||
BC060850.1 | - | 3 | 950 | 4820254 | ||
BC068556.1 | - | 3 | 957 | 4820792 | ||
BX537565.1 | - | 3 | 940 |