Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182922.2 | NP_891552.1 | 9 | 1352 | ||
XM_005256013.2 | XP_005256070.1 | 8 | 1167 | |||
XM_011523188.2 | XP_011521490.1 | 9 | 1242 | |||
XM_017023385.1 | XP_016878874.1 | 7 | 1305 | |||
XM_017023386.1 | XP_016878875.1 | 7 | 1110 | |||
GenBank mRNA | AF462442.1 | - | 7 | 1265 | ||
AK000725.1 | - | 8 | 1159 | |||
AK289949.1 | - | 9 | 1317 | |||
BC014415.1 | - | 1329 | ||||
BC018730.1 | - | 9 | 1317 | 4907047 | ||
BC033077.1 | - | 9 | 1218 | 4301559 |