Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194286.3 | NP_919262.2 | 13 | 6265 | ||
GenBank mRNA | AK125400.1 | - | 1 | 1004 |