Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000328.2 | NP_000319.1 | 11, 12 | 1587 | ||
NM_001034853.1 | NP_001030025.1 | 11, 12 | 1587 | |||
XM_005272633.2 | XP_005272690.1 | 11, 12 | 1616 | |||
XM_011543940.2 | XP_011542242.1 | 11, 12 | 1583 | |||
XM_017029710.1 | XP_016885199.1 | 11, 12 | 1616 | |||
XM_017029711.1 | XP_016885200.1 | 11, 12 | 1613 | |||
XM_017029712.1 | XP_016885201.1 | 11, 12 | 1613 | |||
GenBank mRNA | AJ238395.1 | - | 11, 12 | 1474 | ||
AK223491.1 | - | 11, 12 | 1561 | |||
AK291832.1 | - | 11, 12 | 1594 | |||
KU178312.1 | - | 11, 12 | 1419 | |||
U57629.1 | - | 11, 12 | 1478 | |||
X97668.1 | - | 11, 12 | 1419 |