Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000539.3 | NP_000530.1 | 1 | 312 | ||
GenBank mRNA | BC112104.1 | - | 1 | 304 | 8327572 | |
BC112106.1 | - | 1 | 304 | 8327574 | ||
BX537381.1 | - | 1 | 311 |
Set Membership: |
G-protein coupled receptor |