Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SNRPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278656.1 | NP_001265585.1 | 3 | 726 | ||
NM_004175.4 | NP_004166.1 | 3 | 740 | |||
GenBank mRNA | AK296173.1 | - | 3 | 383 | ||
CN286602.1 | - | 3 | 380 | |||
CR456583.1 | - | 3 | 416 |