Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244974.1 | NP_001231903.1 | 8 | 1462 | ||
NM_002710.3 | NP_002701.1 | 7 | 2362 | |||
GenBank mRNA | AK091841.1 | - | 1 | 2087 | ||
L07395.1 | - | 7 | 2079 | |||
X74008.1 | - | 7 | 2245 |