Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321053.1 | NP_001307982.1 | 3 | 513 | ||
NM_002727.3 | NP_002718.2 | 2 | 298 | |||
XM_017016392.1 | XP_016871881.1 | 3 | 926 | |||
GenBank mRNA | AK311873.1 | - | 2 | 210 | ||
BC015516.1 | - | 2 | 208 | 3883161 | ||
BG548665.1 | - | 2 | 178 | |||
BP335278.1 | - | 2 | 254 | |||
BP363056.1 | - | 2 | 255 | |||
CB110512.1 | - | 2 | 254 | |||
J03223.1 | - | 2 | 199 | |||
X12765.1 | - | 1 | 68 | |||
X17042.1 | - | 2 | 192 |