Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BEST1 SIRNA Assays ›
Gene Symbol:
BEST1
Gene Name:
bestrophin 1
Gene Aliases:
ARB, BEST, BMD, RP50, TU15B, VMD2
Chromosome Location:
Chr.11: 61946722 - 61964465 on Build GRCh38
UniGene ID:
Hs.524910
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14228
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BEST1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001139443.1
NP_001132915.1
9
1614
NM_001300786.1
NP_001287715.1
8
1533
NM_001300787.1
NP_001287716.1
9
1614
NM_004183.3
NP_004174.1
10
1802
NR_134580.1
10
2005
XM_005274210.3
XP_005274267.1
10
1912
XM_005274215.3
XP_005274272.1
8
1567
XM_005274216.3
XP_005274273.1
9
1927
XM_005274219.3
XP_005274276.1
8
1679
XM_005274221.3
XP_005274278.1
7
1526
XM_011545229.2
XP_011543531.1
10
1352
XM_011545230.2
XP_011543532.1
9
1155
XM_011545233.2
XP_011543535.1
4
1049
XM_017018228.1
XP_016873717.1
8
1567
XM_017018229.1
XP_016873718.1
10
2115
XM_017018230.1
XP_016873719.1
8
1770
GenBank mRNA
AB209451.1
-
6
5054
AF057169.1
-
10
1326
AF057170.1
-
10
1529
AF073501.1
-
10
1310
AK289681.1
-
10
1533
AK295580.1
-
7
949
AK295998.1
-
9
1148
AK298747.1
-
5
861
AK301392.1
-
8
1131
AY515704.1
-
9
1222
BC015220.2
-
8
1533
3877806
BC041664.1
-
9
1128
5194649
JQ954696.1
-
9
1288
KU178390.1
-
7
961
KU178391.1
-
8
1042
KU178392.1
-
7
889
KU178393.1
-
3
423
KU178394.1
-
3
267
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chloride transport
visual perception
bicarbonate transport
transepithelial chloride transport
ion transmembrane transport
detection of light stimulus involved in visual perception
regulation of calcium ion transport
chloride transmembrane transport
intracellular calcium activated chloride channel activity
chloride channel activity
bicarbonate transmembrane transporter activity
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0