Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTPRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109754.2 | NP_001103224.1 | 4 | 934 | ||
NM_001206971.1 | NP_001193900.1 | 2 | 265 | |||
NM_001206972.1 | NP_001193901.1 | 2 | 265 | |||
NM_002837.4 | NP_002828.3 | 2 | 265 | |||
XM_006719528.1 | XP_006719591.1 | 4 | 934 | |||
XM_006719529.3 | XP_006719592.1 | 2 | 761 | |||
XM_011538614.1 | XP_011536916.1 | 4 | 934 | |||
XM_017019723.1 | XP_016875212.1 | 4 | 934 | |||
GenBank mRNA | AK126121.1 | - | 4 | 935 | ||
AK128562.1 | - | 4 | 542 | |||
BC051329.1 | - | 4 | 908 | |||
BC101679.1 | - | 2 | 264 | 8069185 | ||
BC113463.1 | - | 2 | 264 | 8322515 | ||
BC143356.1 | - | 2 | 264 | 9051861 | ||
BC143360.1 | - | 2 | 264 | 9051865 | ||
BX647238.1 | - | 4 | 913 | |||
BX648245.1 | - | 4 | 917 | |||
CR749546.1 | - | 4 | 915 | |||
X54131.1 | - | 2 | 265 |