Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DGKB SIRNA Assays ›
Gene Symbol:
DGKB
Gene Name:
diacylglycerol kinase beta
Gene Aliases:
DAGK2, DGK, DGK-BETA
Chromosome Location:
Chr.7: 14145049 - 14974777 on Build GRCh38
UniGene ID:
Hs.567255
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1425
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DGKB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004080.2
NP_004071.1
15, 16
1549
NM_145695.2
NP_663733.1
15, 16
1549
XM_005249628.2
XP_005249685.1
16, 17
2077
XM_005249629.2
XP_005249686.1
16, 17
2074
XM_005249630.2
XP_005249687.1
15, 16
1590
XM_005249631.3
XP_005249688.1
14, 15
1805
XM_011515153.2
XP_011513455.1
16, 17
1915
XM_011515154.2
XP_011513456.1
16, 17
1631
XM_011515156.2
XP_011513458.1
15, 16
1587
XM_011515157.2
XP_011513459.1
15, 16
1594
XM_011515158.2
XP_011513460.1
14, 15
1730
XM_017011783.1
XP_016867272.1
16, 17
1968
XM_017011784.1
XP_016867273.1
16, 17
1628
XM_017011785.1
XP_016867274.1
15, 16
1862
XM_017011786.1
XP_016867275.1
15, 16
1842
XM_017011787.1
XP_016867276.1
15, 16
1592
XM_017011788.1
XP_016867277.1
15, 16
1878
XM_017011789.1
XP_016867278.1
15, 16
1595
XM_017011790.1
XP_016867279.1
16, 17
1914
XM_017011791.1
XP_016867280.1
16, 17
1913
XM_017011792.1
XP_016867281.1
16, 17
1913
GenBank mRNA
AB018261.1
-
12, 13
1178
AK304719.1
-
14, 15
1805
BC105005.1
-
15, 16
1429
8144008
BC143632.1
-
15, 16
1426
9052145
BC143633.1
-
14, 15
1393
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_019304, XM_008764650, XM_008764649, XM_008764651, D16100.This siRNA also targets the following transcripts in Mouse: NM_178681, AK139408, AK122355, BC070461, AK032509.
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
platelet activation
intracellular signal transduction
diacylglycerol kinase activity
calcium ion binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0