Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KYAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122671.1 | NP_001116143.1 | 13 | 1531 | ||
NM_001122672.1 | NP_001116144.1 | 12 | 1387 | |||
NM_001287390.1 | NP_001274319.1 | 15 | 1928 | |||
NM_004059.4 | NP_004050.3 | 13 | 1537 | |||
NR_109829.1 | 13 | 1569 | ||||
XM_006717311.3 | XP_006717374.1 | 14 | 1723 | |||
XM_011519165.2 | XP_011517467.1 | 15 | 1899 | |||
XM_011519167.2 | XP_011517469.2 | 13 | 1503 | |||
XM_011519168.1 | XP_011517470.1 | 12 | 1362 | |||
XM_011519169.1 | XP_011517471.1 | 13 | 1631 | |||
XM_011519170.2 | XP_011517472.1 | 13 | 1999 | |||
XM_017015260.1 | XP_016870749.1 | 14 | 1683 | |||
XM_017015261.1 | XP_016870750.1 | 15 | 1905 | |||
XM_017015262.1 | XP_016870751.1 | 15 | 2394 | |||
XM_017015263.1 | XP_016870752.1 | 15 | 1809 | |||
XM_017015264.1 | XP_016870753.1 | 14 | 1716 | |||
XM_017015265.1 | XP_016870754.1 | 13 | 2093 | |||
XM_017015266.1 | XP_016870755.1 | 14 | 1724 | |||
XM_017015267.1 | XP_016870756.1 | 13 | 1545 | |||
XM_017015268.1 | XP_016870757.1 | 14 | 1707 | |||
GenBank mRNA | AK094505.1 | - | 13 | 2112 | ||
AK291178.1 | - | 13 | 1525 | |||
AK297995.1 | - | 15 | 1784 | |||
BC021262.2 | - | 13 | 1530 | 4137942 | ||
BC033685.1 | - | 12 | 1241 | 5193675 |