Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039573.2 | NP_001034662.2 | 9 | 1249 | ||
NM_001143998.1 | NP_001137470.1 | 9 | 1249 | |||
NM_001143999.1 | NP_001137471.1 | 9 | 1245 | |||
NM_001144001.1 | NP_001137473.1 | 7 | 1037 | |||
NM_001204408.1 | NP_001191337.1 | 11 | 1594 | |||
NM_001204410.1 | NP_001191339.1 | 10 | 1287 | |||
NM_003003.3 | NP_002994.3 | 9 | 1196 | |||
GenBank mRNA | AB208811.1 | - | 9 | 1228 | ||
AK290913.1 | - | 9 | 1142 | |||
AK293206.1 | - | 7 | 1037 | |||
BC136523.1 | - | 9 | 1084 | 9020513 | ||
BC136525.1 | - | 9 | 1084 | 9020515 | ||
BC142979.1 | - | 9 | 1140 | 8860414 | ||
BX537448.1 | - | 9 | 1150 | |||
D67029.1 | - | 9 | 1232 | |||
FM995495.1 | - | 11 | 1488 | |||
FM995496.1 | - | 10 | 1285 | |||
FM995497.1 | - | 10 | 1384 |