Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003019.4 | NP_003010.4 | 8 | 1036 | ||
XM_011540087.1 | XP_011538389.1 | 8 | 1015 | |||
XM_011540088.2 | XP_011538390.1 | 7 | 987 | |||
GenBank mRNA | BC022318.1 | - | 8 | 1037 | 4696173 | |
CR541948.1 | - | 7 | 993 | |||
X65018.1 | - | 8 | 1164 |