Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003061.2 | NP_003052.2 | 34 | 3979 | ||
GenBank mRNA | AB011537.3 | - | 34 | 3979 | ||
AB017167.1 | - | 34 | 3959 | |||
BC146761.1 | - | 34 | 3979 | 9007200 | ||
BC146851.1 | - | 34 | 3892 | 9056779 |