Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271946.1 | NP_001258875.1 | 4, 5 | 830 | ||
NM_003062.3 | NP_003053.1 | 4, 5 | 830 | |||
XM_017009779.1 | XP_016865268.1 | 4, 5 | 233 | |||
GenBank mRNA | AB011538.2 | - | 4, 5 | 830 | ||
AB017169.1 | - | 4, 5 | 673 | |||
AK310549.1 | - | 4, 5 | 447 | |||
AY358884.1 | - | 4, 5 | 492 | |||
BC146759.1 | - | 4, 5 | 830 | 9007197 |