Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301069.1 | NP_001287998.1 | 6 | 887 | ||
NM_003089.5 | NP_003080.2 | 6 | 887 | |||
GenBank mRNA | AK096783.1 | - | 6 | 576 | ||
AK293120.1 | - | 6 | 606 | |||
AL560281.3 | - | 6 | 575 | |||
BC001315.1 | - | 6 | 560 | 3454479 | ||
M22636.1 | - | 6 | 551 | |||
X06812.1 | - | 4 | 235 | |||
X06815.1 | - | 6 | 531 | |||
X84841.1 | - | 5 | 380 |