Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003126.2 | NP_003117.2 | 2 | 437 | ||
XM_011509916.2 | XP_011508218.1 | 3 | 315 | |||
XM_011509917.2 | XP_011508219.1 | 2 | 351 | |||
XM_011509918.2 | XP_011508220.1 | 2 | 351 | |||
XM_011509919.2 | XP_011508221.1 | 2 | 353 | |||
GenBank mRNA | M61877.1 | - | 2 | 424 |