Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004106.1 | NP_004097.1 | 5 | 252 | ||
GenBank mRNA | AK307102.1 | - | 5 | 277 | ||
BC033872.1 | - | 5 | 249 | 5219148 | ||
M33195.1 | - | 5 | 252 |